Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077116.2 | NP_001070584.1 | 1 - 2 | 83 | 70 | |
GenBank mRNA | BC102605.1 | - | 1 - 2 | 86 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |