Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077422.3 | NP_001070890.2 | 2 - 3 | 340 | 122 | |
GenBank mRNA | BC102940.1 | - | 2 - 3 | 341 | 122 | |
BC133477.1 | - | 2 - 3 | 340 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |