Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077944.2 | NP_001071412.1 | 1 - 2 | 46 | 177 | |
GenBank mRNA | BC111609.1 | - | 1 - 2 | 47 | 177 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |