Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174131.4 | NP_776556.3 | 1 - 2 | 467 | 74 | |
GenBank mRNA | BC102630.1 | - | 1 - 2 | 467 | 74 | |
DT852369.1 | - | 1 - 2 | 458 | 74 | ||
X69514.1 | - | 358 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |