Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174345.4 | NP_776770.2 | 3 - 4 | 487 | 90 | |
GenBank mRNA | BC105182.1 | - | 3 - 4 | 488 | 90 | |
BT030487.1 | - | 3 - 4 | 473 | 90 | ||
X53335.1 | - | 412 | 90 | |||
X53827.1 | - | 446 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |