Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174724.4 | NP_777149.1 | 185 | 88 | ||
GenBank mRNA | BC104564.1 | - | 186 | 88 | ||
BT025455.1 | - | 345 | 88 | |||
X06089.1 | - | 135 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus |