Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JCHAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175773.3 | NP_786967.1 | 1 - 2 | 91 | 91 | |
GenBank mRNA | BC103426.1 | - | 1 - 2 | 94 | 91 | |
L20311.1 | - | 1 - 2 | 64 | 91 | ||
U02301.1 | - | 1 - 2 | 64 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |