Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175813.2 | NP_787007.1 | 6 - 7 | 863 | 153 | |
GenBank mRNA | BC104590.1 | - | 6 - 7 | 863 | 153 | |
X53689.1 | - | 632 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |