Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSF2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035493.3 | NP_001030570.1 | 7 - 8 | 1009 | 67 | |
GenBank mRNA | BC110166.1 | - | 7 - 8 | 1010 | 67 | |
BT021887.1 | - | 7 - 8 | 1022 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |