Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GID8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037587.2 | NP_001032676.1 | 4 - 5 | 587 | 62 | |
GenBank mRNA | BC109762.1 | - | 4 - 5 | 588 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |