Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037614.2 | NP_001032703.1 | 1 - 2 | 503 | 71 | |
GenBank mRNA | BC109502.1 | - | 1 - 2 | 504 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |