Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELPLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037628.2 | NP_001032717.2 | 1 - 2 | 131 | 57 | |
GenBank mRNA | AY929857.2 | - | 1 - 2 | 87 | 57 | |
BC140648.1 | - | 1 - 2 | 132 | 57 | ||
EV613550.1 | - | 1 - 2 | 60 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |