Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037816.1 | NP_001032905.1 | 5 - 6 | 829 | 60 | |
GenBank mRNA | BC111139.1 | - | 5 - 6 | 829 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |