Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H11orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038037.2 | NP_001033126.1 | 6 - 7 | 696 | 75 | |
GenBank mRNA | BC109973.1 | - | 6 - 7 | 697 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |