Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038050.2 | NP_001033139.1 | 3 - 4 | 360 | 87 | |
GenBank mRNA | BC111178.1 | - | 3 - 4 | 361 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |