Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNSTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038055.2 | NP_001033144.1 | 7 - 8 | 875 | 123 | |
GenBank mRNA | BC111170.1 | - | 7 - 8 | 876 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |