Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038060.1 | NP_001033149.1 | 3 - 4 | 1732 | 70 | |
GenBank mRNA | BC111157.1 | - | 3 - 4 | 1732 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |