Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038063.2 | NP_001033152.1 | 1 - 2 | 307 | 58 | |
GenBank mRNA | BC111266.1 | - | 1 - 2 | 308 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |