Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038075.1 | NP_001033164.1 | 5 - 6 | 549 | 125 | |
GenBank mRNA | BC110030.1 | - | 5 - 6 | 549 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |