Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038090.2 | NP_001033179.1 | 6 - 7 | 916 | 62 | |
GenBank mRNA | BC109917.1 | - | 6 - 7 | 917 | 62 | |
BC148921.1 | - | 5 - 6 | 1465 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |