Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038111.2 | NP_001033200.1 | 1 - 2 | 473 | 61 | |
GenBank mRNA | BC109918.1 | - | 1 - 2 | 474 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |