Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C27H4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038136.2 | NP_001033225.1 | 4 - 5 | 848 | 91 | |
GenBank mRNA | BC111359.1 | - | 4 - 5 | 849 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |