Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C27H4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038136.2 | NP_001033225.1 | 6 - 7 | 1097 | 86 | |
GenBank mRNA | BC111359.1 | - | 6 - 7 | 1098 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |