Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038138.2 | NP_001033227.1 | 1 - 2 | 256 | 76 | |
GenBank mRNA | BC110001.1 | - | 1 - 2 | 257 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |