Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038138.2 | NP_001033227.1 | 3 - 4 | 601 | 82 | |
GenBank mRNA | BC110001.1 | - | 3 - 4 | 602 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |