Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038164.2 | NP_001033253.1 | 1 - 2 | 31 | 110 | |
GenBank mRNA | BC111171.1 | - | 1 - 2 | 32 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |