Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038166.2 | NP_001033255.1 | 4 - 5 | 794 | 108 | |
GenBank mRNA | BC111168.1 | - | 4 - 5 | 795 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |