Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038182.2 | NP_001033271.1 | 4 - 5 | 510 | 86 | |
GenBank mRNA | BC109620.1 | - | 4 - 5 | 511 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |