Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIBC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038191.2 | NP_001033280.2 | 5 - 6 | 1072 | 77 | |
GenBank mRNA | BC109543.1 | - | 5 - 6 | 1073 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |