Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038203.1 | NP_001033292.1 | 10 - 11 | 1194 | 94 | |
GenBank mRNA | BC109852.1 | - | 10 - 11 | 1194 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |