Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038223.2 | NP_001033312.1 | 6 - 7 | 549 | 105 | |
GenBank mRNA | BC109806.1 | - | 6 - 7 | 550 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |