Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038509.2 | NP_001033598.1 | 2 - 3 | 176 | 142 | |
GenBank mRNA | BC110005.1 | - | 2 - 3 | 178 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |