Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038512.2 | NP_001033601.1 | 2 - 3 | 1380 | 114 | |
GenBank mRNA | BC109844.1 | - | 2 - 3 | 1381 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |