Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038512.2 | NP_001033601.1 | 6 - 7 | 1672 | 126 | |
GenBank mRNA | BC109844.1 | - | 6 - 7 | 1673 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |