Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038514.2 | NP_001033603.1 | 5 - 6 | 725 | 61 | |
GenBank mRNA | BC109750.1 | - | 5 - 6 | 726 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |