Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038520.2 | NP_001033609.1 | 4 - 5 | 364 | 108 | |
GenBank mRNA | BC109885.1 | - | 4 - 5 | 366 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |