Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECHDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038536.2 | NP_001033625.1 | 6 - 7 | 551 | 124 | |
GenBank mRNA | BC109686.1 | - | 6 - 7 | 552 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |