Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038686.2 | NP_001033775.1 | 2 - 3 | 149 | 118 | |
GenBank mRNA | BC110230.1 | - | 2 - 3 | 150 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |