Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038688.2 | NP_001033777.1 | 1 - 2 | 170 | 81 | |
GenBank mRNA | BC110229.1 | - | 1 - 2 | 171 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |