Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040499.2 | NP_001035589.1 | 4 - 5 | 732 | 126 | |
GenBank mRNA | BC110019.1 | - | 4 - 5 | 735 | 126 | |
BT025411.1 | - | 2 - 3 | 297 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |