Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040607.1 | NP_001035697.1 | 1 - 2 | 874 | 122 | |
GenBank mRNA | BC116068.1 | - | 1 - 2 | 861 | 122 | |
DQ333219.1 | - | 1 - 2 | 874 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |