Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046031.2 | NP_001039496.1 | 6 - 7 | 704 | 86 | |
GenBank mRNA | BC110244.1 | - | 6 - 7 | 705 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |