Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046059.2 | NP_001039524.1 | 2 - 3 | 268 | 77 | |
GenBank mRNA | BC111670.1 | - | 2 - 3 | 269 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |