Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046104.2 | NP_001039569.1 | 8 - 9 | 1084 | 80 | |
GenBank mRNA | BC112535.1 | - | 8 - 9 | 1085 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |