Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM207A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046108.2 | NP_001039573.1 | 4 - 5 | 535 | 111 | |
GenBank mRNA | BC112883.1 | - | 4 - 5 | 536 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |