Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046149.1 | NP_001039614.1 | 3 - 4 | 433 | 75 | |
GenBank mRNA | BC105336.1 | - | 3 - 4 | 433 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |