Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046186.2 | NP_001039651.1 | 2 - 3 | 624 | 54 | |
GenBank mRNA | BC112464.1 | - | 2 - 3 | 625 | 54 | |
BT029911.1 | - | 3 - 4 | 647 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |