Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046259.2 | NP_001039724.1 | 1 - 2 | 659 | 86 | |
GenBank mRNA | BC110202.1 | - | 1 - 2 | 660 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |