Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046390.2 | NP_001039855.1 | 1 - 2 | 189 | 79 | |
GenBank mRNA | BC110254.1 | - | 1 - 2 | 190 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |