Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046438.1 | NP_001039903.1 | 2 - 3 | 433 | 91 | |
GenBank mRNA | BC112687.1 | - | 2 - 3 | 433 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |